1-Chloro-4-(4-iodophenoxy)benzene 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloro-4'-iododiphenyl ether
OR11362-1G
OR11362-5G
APOSOR11362-1GEA
2650
CZK
APOSOR11362-1G
APOSOR11362-5G
1-Chloro-4-(4-iodophenoxy)benzene 99%
1-Chloro-4-(4-iodophenoxy)benzene
Vzorec:
C₁₂H₈ClIO Bod tání: 60…63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06796299 CAS číslo: 854257-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...