Tisk…

4-Amino-4'-methyldiphenyl ether

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(p-Tolyloxy)aniline, 4-Aminophenyl p-Tolyl Ether 4-(p-Tolyloxy)aniline, 4-Amino-4'-methyldiphenyl Ether, 4-Aminophenyl p-Tolyl Ether

OR11354-25G OR11354-5G OR11354-100G
APOSOR11354-25GEA 2480 CZK
APOSOR11354-25G APOSOR11354-5G APOSOR11354-100G
4-Amino-4'-methyldiphenyl ether
4-Amino-4'-methyldiphenyl ether
Vzorec: C₁₃H₁₃NO
MW: 199,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059858
CAS číslo: 41295-20-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR