4-Chloro-1-naphthol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloronaphthalen-1-ol, 1-Chloro-4-hydroxynaphthalene, 4-CN, Chloronaphthol
OR11309-25G
OR11309-10G
OR11309-100G
APOSOR11309-25GEA
3740
CZK
APOSOR11309-25G
APOSOR11309-10G
APOSOR11309-100G
4-Chloro-1-naphthol
4-Chloro-1-naphthol
Vzorec:
ClC₁₀H₆OH MW: 178,62 g/mol Bod tání: 120…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003974 CAS číslo: 604-44-4 EINECS: 210-068-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...