Tisk…

2-Amino-4,6-dihydroxypyrimidine 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-6-hydroxy-1H-pyrimidin-4-one, 2-Amino-6-hydroxypyrimidin-4(3H)-one

OR11291-500G
APOSOR11291-500GEA 2530 CZK
APOSOR11291-500G
2-Amino-4,6-dihydroxypyrimidine 95%
2-Amino-4,6-dihydroxypyrimidine
Vzorec: C₄H₅N₃O₂
MW: 127,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006094
CAS číslo: 56-09-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR