Tisk…

ω-Bromo-3-hydroxyacetophenone 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: α-Bromo-3-hydroxyacetophenone, 3-Hydroxyphenacyl bromide, 2-Bromo-1-(3-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(3-hydroxyphenyl)ethanone, 2-Bromo-3'-hydroxyacetophenone

OR11288-1G OR11288-25G OR11288-5G
APOSOR11288-1GEA 622 CZK
APOSOR11288-1G APOSOR11288-25G APOSOR11288-5G
ω-Bromo-3-hydroxyacetophenone 96%
ω-Bromo-3-hydroxyacetophenone
Vzorec: C₈H₇BrO₂
Bod tání: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD05664326
CAS číslo: 2491-37-4
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR