2'-Methylacetophenone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
o-Methylacetophenone, Methyl o-tolyl ketone, 1-(o-Tolyl)ethanone
OR11271-100G
OR11271-25G
OR11271-500G
APOSOR11271-100GEA
953
CZK
APOSOR11271-100G
APOSOR11271-25G
APOSOR11271-500G
2'-Methylacetophenone
2'-Methylacetophenone
Vzorec:
C₉H₁₀O MW: 134.18 Bod varu: 214 °C Hustota: 1.026 g/cm³ (25 °C) Bod vzplanutí: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008734 CAS číslo: 577-16-2 EINECS: 209-408-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...