(1-Naphthoxy)acetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Naphthalen-1-yloxy)acetic acid
OR11263-100G
OR11263-25G
APOSOR11263-100GEA
15680
CZK
APOSOR11263-100G
APOSOR11263-25G
(1-Naphthoxy)acetic acid 98%
(1-Naphthoxy)acetic acid
Vzorec:
C₁₂H₁₀O₃ MW: 202,21 g/mol Bod tání: 193…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003927 CAS číslo: 2976-75-2 EINECS: 221-023-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...