Trithiocyanuric acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR11248-100G
OR11248-500G
APOSOR11248-100GEA
1660
CZK
APOSOR11248-100G
APOSOR11248-500G
Trithiocyanuric acid 98%
Trithiocyanuric acid
Vzorec:
C₃H₃N₃S₃ Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006052 CAS číslo: 638-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...