1-Naftylizokyanát
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Naphthylisocyanate
OR11220-100G
OR11220-250G
OR11220-25G
APOSOR11220-100GEA
2240
CZK
APOSOR11220-100G
APOSOR11220-250G
APOSOR11220-25G
1-Naftylizokyanát
1-Naftylizokyanát
- May form small amount of sediment.
Vzorec:
C₁₁H₇NO MW: 169,18 g/mol Bod varu: 267 °C Bod tání: 4 °C Hustota: 1,177 g/cm³ (25 °C) Bod vzplanutí: 113 °C Storage Temperature: Freezer |
MDL Number:
MFCD00003881 CAS číslo: 86-84-0 EINECS: 201-703-7 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...