Tisk…

3-Hydroxy-o-toluic acid 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Hydroxy-2-methylbenzoic acid

OR11194-25G OR11194-100G
APOSOR11194-25GEA 622 CZK
APOSOR11194-25G APOSOR11194-100G
3-Hydroxy-o-toluic acid 99%
3-Hydroxy-o-toluic acid
Vzorec: C₈H₈O₃
MW: 152,15 g/mol
Bod tání: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00671541
CAS číslo: 603-80-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR