3,5-Dimethylbenzhydrazide
Dodavatel: Apollo Scientific
Synonyms:
3,5-Dimethylbenzohydrazide
OR111592-1G
OR111592-5G
APOSOR111592-1GEA
2480
CZK
APOSOR111592-1G
APOSOR111592-5G
3,5-Dimethylbenzhydrazide
3,5-Dimethylbenzhydrazide
Vzorec:
C₉H₁₂N₂O MW: 164,204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01993717 CAS číslo: 27389-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...