Tisk…

3-Amino-1-ethyl-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Ethyl-1H-pyrazol-3-amine

OR111552-10G OR111552-1G OR111552-5G
APOSOR111552-10GEA 16340 CZK
APOSOR111552-10G APOSOR111552-1G APOSOR111552-5G
3-Amino-1-ethyl-1H-pyrazole
3-Amino-1-ethyl-1H-pyrazole
Vzorec: C₅H₉N₃
MW: 111,1451 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03422571
CAS číslo: 55361-49-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR