Tisk…

4-Methyl-5-nitroimidazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 1H-Imidazole, 4-methyl-5-nitro-, 5-methyl-4-nitro-1H-imidazole, 4-METHYL-5-NITRO-1H-IMIDAZOLE

OR111542-1G OR111542-25G OR111542-5G
APOSOR111542-1GEA 622 CZK
APOSOR111542-1G APOSOR111542-25G APOSOR111542-5G
4-Methyl-5-nitroimidazole
4-Methyl-5-nitroimidazole
Vzorec: C₄H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01721395
CAS číslo: 14003-66-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR