5,6-Dichloro-1H-1,3-benzimidazole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5,6-Dichlorobenzimidazole, 5,6-Dichloro-1H-benzo[d]imidazole
OR11150-1G
OR11150-250MG
APOSOR11150-1GEA
1450
CZK
APOSOR11150-1G
APOSOR11150-250MG
5,6-Dichloro-1H-1,3-benzimidazole
5,6-Dichloro-1H-1,3-benzimidazole
Vzorec:
C₇H₄Cl₂N₂ MW: 187,03 g/mol Bod tání: 205…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00211325 CAS číslo: 6478-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...