5,6-Dihydro-4H-cyclopenta[d][1,3]thiazol-2-amine
Dodavatel: Apollo Scientific
Synonyms:
2-Amino-5,6-dihydro-4H-cyclopenta[d]thiazole
OR111480-1G
OR111480-5G
APOSOR111480-1GEA
0
CZK
APOSOR111480-1G
APOSOR111480-5G
5,6-Dihydro-4H-cyclopenta[d][1,3]thiazol-2-amine
5,6-Dihydro-4H-cyclopenta[d][1,3]thiazol-2-amine
Vzorec:
C₆H₈N₂S MW: 140,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00159774 CAS číslo: 53051-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...