Tisk…

6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Cyclopropyl-2-hydroxynicotinonitrile

OR111473-1G OR111473-250MG
APOSOR111473-1GEA 1370 CZK
APOSOR111473-1G APOSOR111473-250MG
6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
6-Cyclopropyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Vzorec: C₉H₈N₂O
MW: 160,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11553070
CAS číslo: 847144-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR