Tisk…

4-Cyanophenyl isocyanate

Dodavatel: Apollo Scientific
Varování

OR11147-250MG OR11147-500MG
APOSOR11147-250MGEA 1660 CZK
APOSOR11147-250MG APOSOR11147-500MG
4-Cyanophenyl isocyanate
4-Cyanophenyl isocyanate

When melted, may contain small amount of sediment.

Vzorec: C₈H₄N₂O
MW: 144,13 g/mol
Bod tání: 101…104 °C
Storage Temperature: Refrigerator
CAS číslo: 40465-45-0
EINECS: 254-934-0
UN: 2206
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR