3,5-Dichloro-4-hydroxybenzyl alcohol
Dodavatel: Apollo Scientific
Synonyms:
2,6-Dichloro-4-(hydroxymethyl)phenol
OR111469-1G
OR111469-250MG
OR111469-500MG
OR111469-2.5G
APOSOR111469-1GEA
13030
CZK
APOSOR111469-1G
APOSOR111469-250MG
APOSOR111469-500MG
APOSOR111469-2.5G
3,5-Dichloro-4-hydroxybenzyl alcohol
3,5-Dichloro-4-hydroxybenzyl alcohol
Vzorec:
C₇H₆Cl₂O₂ MW: 193,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06202847 CAS číslo: 22002-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...