Tisk…

3-(tert-Butoxycarbonyl)-1,3-thiazinane-4-carboxylic acid

Dodavatel: Apollo Scientific

OR111421-1G OR111421-250MG OR111421-500MG
APOSOR111421-1GEA 15940 CZK
APOSOR111421-1G APOSOR111421-250MG APOSOR111421-500MG
3-(tert-Butoxycarbonyl)-1,3-thiazinane-4-carboxylic acid
3-(tert-Butoxycarbonyl)-1,3-thiazinane-4-carboxylic acid
Vzorec: C₁₀H₁₇NO₄S
Storage Temperature: Ambient
MDL Number: MFCD26099141
CAS číslo: 590409-14-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR