3-Amino-3-(2-bromophenyl)propionic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR111400-10G
OR111400-5G
OR111400-1G
APOSOR111400-10GEA
2530
CZK
APOSOR111400-10G
APOSOR111400-5G
APOSOR111400-1G
3-Amino-3-(2-bromophenyl)propionic acid
3-Amino-3-(2-bromophenyl)propionic acid
Vzorec:
C₉H₁₀BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01871297 CAS číslo: 117391-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...