Tisk…

α-Iodo-4-nitrotoluene

Dodavatel: Apollo Scientific
Varování

OR111344-1G OR111344-5G
APOSOR111344-1GEA 3230 CZK
APOSOR111344-1G APOSOR111344-5G
α-Iodo-4-nitrotoluene
α-Iodo-4-nitrotoluene
Vzorec: C₇H₆INO₂
Storage Temperature: Ambient
CAS číslo: 3145-86-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR