4,5,6,7-Tetrahydro-benzo[b]thiophene-2-carbonyl chloride
Dodavatel: Apollo Scientific
OR111333-10G
OR111333-1G
OR111333-5G
APOSOR111333-10GEA
0
CZK
APOSOR111333-10G
APOSOR111333-1G
APOSOR111333-5G
4,5,6,7-Tetrahydro-benzo[b]thiophene-2-carbonyl chloride
4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carbonyl chloride
Vzorec:
C₉H₉ClOS Storage Temperature: Ambient |
MDL Number:
MFCD04968788 CAS číslo: 65361-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...