Tisk…

Benzamidine hydrochloride hydrate

Dodavatel: Apollo Scientific
Varování

OR11123-100G OR11123-25G OR11123-5G
APOSOR11123-100GEA 0 CZK
APOSOR11123-100G APOSOR11123-25G APOSOR11123-5G
Benzamidine hydrochloride hydrate
Benzamidine hydrochloride hydrate
Vzorec: C₆H₅C(=NH)NH₂·HCl·H₂O
Bod tání: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066285
CAS číslo: 206752-36-5
EINECS: 216-795-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR