3'-Bromo-5'-chloro-2'-hydroxyacetophenone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(3-Bromo-5-chloro-2-hydroxyphenyl)ethanone, 5-Chloro-3-bromo-2-hydroxyacetophenone
OR1112-25G
OR1112-5G
APOSOR1112-25GEA
1990
CZK
APOSOR1112-25G
APOSOR1112-5G
3'-Bromo-5'-chloro-2'-hydroxyacetophenone
3'-Bromo-5'-chloro-2'-hydroxyacetophenone
Vzorec:
C₈H₆BrClO₂ MW: 249,49 g/mol Bod tání: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00857068 CAS číslo: 59443-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...