Tisk…

(4-Aminophenyl)(morpholino)methanone

Dodavatel: Apollo Scientific

OR111157-1G OR111157-5G
APOSOR111157-1GEA 2610 CZK
APOSOR111157-1G APOSOR111157-5G
(4-Aminophenyl)(morpholino)methanone
(4-Aminophenyl)(morpholino)methanone
Vzorec: C₁₁H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00158788
CAS číslo: 51207-86-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR