Tisk…

3-(4-Methoxyphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid

Dodavatel: Apollo Scientific

OR111110-1G OR111110-5G
APOSOR111110-1GEA 0 CZK
APOSOR111110-1G APOSOR111110-5G
3-(4-Methoxyphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
3-(4-Methoxyphenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
Vzorec: C₁₈H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD02571654
CAS číslo: 167886-73-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR