Tisk…

Ethyl-3-bromoimidazo[1,2-a]pyrimidine-2-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 3-bromoimidazo[1,2-a]pyrimidine-2-carboxylate

OR111095-1G OR111095-5G OR111095-250MG
APOSOR111095-1GEA 5100 CZK
APOSOR111095-1G APOSOR111095-5G APOSOR111095-250MG
Ethyl-3-bromoimidazo[1,2-a]pyrimidine-2-carboxylate
Ethyl-3-bromoimidazo[1,2-a]pyrimidine-2-carboxylate
Vzorec: C₉H₈BrN₃O₂
MW: 270,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02186429
CAS číslo: 134044-63-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR