Tisk…

3-Bromo-1H-indole-2-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromoindole-2-carboxylic Acid

OR111021-1G OR111021-5G OR111021-25G
APOSOR111021-1GEA 1290 CZK
APOSOR111021-1G APOSOR111021-5G APOSOR111021-25G
3-Bromo-1H-indole-2-carboxylic acid
3-Bromo-1H-indole-2-carboxylic acid
Vzorec: C₉H₆BrNO₂
MW: 240,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02710285
CAS číslo: 28737-33-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR