Tisk…

4-(4-Nitrobenzyl)morpholine

Dodavatel: Apollo Scientific

OR110998-1G OR110998-5G
APOSOR110998-1GEA 0 CZK
APOSOR110998-1G APOSOR110998-5G
4-(4-Nitrobenzyl)morpholine
4-(4-Nitrobenzyl)morpholine
Vzorec: C₁₁H₁₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00448629
CAS číslo: 6425-46-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR