Tisk…

2-Acetyl-5-bromobenzofuran

Dodavatel: Apollo Scientific
Varování

OR110915-1G OR110915-5G
APOSOR110915-1GEA 1450 CZK
APOSOR110915-1G APOSOR110915-5G
2-Acetyl-5-bromobenzofuran
2-Acetyl-5-bromobenzofuran
Vzorec: C₁₀H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD00276737
CAS číslo: 38220-75-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR