1-Bromo-5-chloro-2-iodobenzene 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR11085-100G
OR11085-25G
OR11085-5G
APOSOR11085-100GEA
3640
CZK
APOSOR11085-100G
APOSOR11085-25G
APOSOR11085-5G
1-Bromo-5-chloro-2-iodobenzene 98%
1-Bromo-5-chloro-2-iodobenzene
Vzorec:
C₆H₃BrClI MW: 190.45 Storage Temperature: Ambient |
MDL Number:
MFCD00672944 CAS číslo: 31928-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...