Ethyl-1H-indazole-4-carboxylate 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 1H-indazole-4-carboxylate
OR110817-1G
OR110817-5G
APOSOR110817-1GEA
1740
CZK
APOSOR110817-1G
APOSOR110817-5G
Ethyl-1H-indazole-4-carboxylate 95%
Ethyl-1H-indazole-4-carboxylate
Vzorec:
C₁₀H₁₀N₂O₂ MW: 190,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07371611 CAS číslo: 885279-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...