Tisk…

(2,4-Dichlorophenoxy)acetic acid hydrazide

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(2,4-Dichlorophenoxy)acetohydrazide, (2,4-Dichlorophenoxy)acetohydrazide, (2,4-Dichlorophenoxy)acetic hydrazide

OR11078-25G
APOSOR11078-25GEA 5390 CZK
APOSOR11078-25G
(2,4-Dichlorophenoxy)acetic acid hydrazide
(2,4-Dichlorophenoxy)acetic acid hydrazide
Vzorec: (C₆H₃Cl₂)OCH₂COONHNH₂
MW: 235,07 g/mol
Bod tání: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00051972
CAS číslo: 28236-62-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR