Tisk…

3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR110747-1G OR110747-250MG
APOSOR110747-1GEA 6190 CZK
APOSOR110747-1G APOSOR110747-250MG
3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Vzorec: C₈H₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02182241
CAS číslo: 354548-73-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR