Tisk…

5-Aminobenzimidazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Amino-1H-benzimidazole

OR110728-1G OR110728-5G
APOSOR110728-1GEA 622 CZK
APOSOR110728-1G APOSOR110728-5G
5-Aminobenzimidazole
5-Aminobenzimidazole
Vzorec: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD00465258
CAS číslo: 934-22-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR