Tisk…

4-Chloro-6-piperidin-1-ylpyrimidin-2-ylamine

Dodavatel: Apollo Scientific
Varování

OR110706-1G OR110706-250MG OR110706-5G
APOSOR110706-1GEA 6620 CZK
APOSOR110706-1G APOSOR110706-250MG APOSOR110706-5G
4-Chloro-6-piperidin-1-ylpyrimidin-2-ylamine
4-Chloro-6-piperidin-1-ylpyrimidin-2-ylamine
Vzorec: C₉H₁₃ClN₄
MW: 212,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664698
CAS číslo: 104637-64-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR