Tisk…

1H-Benzimidazole-2-sulphonic acid

Dodavatel: Apollo Scientific

Synonyms: 1H-Benzo[d]imidazole-2-sulphonic acid

OR110696-5G
APOSOR110696-5GEA 3610 CZK
APOSOR110696-5G
1H-Benzimidazole-2-sulphonic acid
1H-Benzimidazole-2-sulphonic acid
Vzorec: C₇H₆N₂O₃S
MW: 198.1991 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100972
CAS číslo: 40828-54-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR