Tisk…

Methyl-2,3-dioxo-1,2,3,4-tetrahydro-6-quinoxalinecarboxylate

Dodavatel: Apollo Scientific

Synonyms: Methyl 2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carboxylate

OR110597-1G
APOSOR110597-1GEA 4040 CZK
APOSOR110597-1G
Methyl-2,3-dioxo-1,2,3,4-tetrahydro-6-quinoxalinecarboxylate
Methyl-2,3-dioxo-1,2,3,4-tetrahydro-6-quinoxalinecarboxylate
Vzorec: C₁₀H₈N₂O₄
MW: 220,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11973607
CAS číslo: 354793-04-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR