Tetrahydro-2H-pyran-4-yl methanesulphonate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Tetrahydropyranyl methanesulphonate
OR110580-1G
OR110580-5G
APOSOR110580-1GEA
622
CZK
APOSOR110580-1G
APOSOR110580-5G
Tetrahydro-2H-pyran-4-yl methanesulphonate
Tetrahydro-2H-pyran-4-yl methanesulphonate
Vzorec:
C₆H₁₂O₄S MW: 180.218 g/mol Bod tání: 46…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD11505063 CAS číslo: 134419-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...