4-bromo-1-methyl-1H-pyrazole-3-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile
OR110567-1G
OR110567-250MG
APOSOR110567-1GEA
3310
CZK
APOSOR110567-1G
APOSOR110567-250MG
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile
Vzorec:
C₅H₄BrN₃ MW: 186,012 g/mol Bod tání: 103…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00103545 CAS číslo: 287922-71-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...