Tisk…

2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-tert-Butylphenoxy)benzaldehyde

OR110496-1G OR110496-5G
APOSOR110496-1GEA 3540 CZK
APOSOR110496-1G APOSOR110496-5G
2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde
2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde
Vzorec: C₁₇H₁₈O₂
MW: 254,33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01568764
CAS číslo: 181297-82-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR