Tisk…

2-(3,5-Dimethylphenoxy)benzaldehyde

Dodavatel: Apollo Scientific
Varování

OR110495-5G OR110495-1G
APOSOR110495-5GEA 0 CZK
APOSOR110495-5G APOSOR110495-1G
2-(3,5-Dimethylphenoxy)benzaldehyde
2-(3,5-Dimethylphenoxy)benzaldehyde
Vzorec: C₁₅H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD01568762
CAS číslo: 320423-51-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR