Tisk…

3-Propoxybenzaldehyde

Dodavatel: Apollo Scientific

OR110480-1G OR110480-5G
APOSOR110480-1GEA 3380 CZK
APOSOR110480-1G APOSOR110480-5G
3-Propoxybenzaldehyde
3-Propoxybenzaldehyde
Vzorec: C₁₀H₁₂O₂
MW: 164.2
Storage Temperature: Ambient
MDL Number: MFCD01993679
CAS číslo: 67698-61-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR