Tisk…

3-Methyl-5-(2-thienyl)-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

OR110456-1G OR110456-5G
APOSOR110456-1GEA 2650 CZK
APOSOR110456-1G APOSOR110456-5G
3-Methyl-5-(2-thienyl)-1H-pyrazole
3-Methyl-5-(2-thienyl)-1H-pyrazole
Vzorec: C₈H₈N₂S
MW: 164,23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00514532
CAS číslo: 31618-80-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR