Methyl 6-hydroxy-5-iodonicotinate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 6-hydroxy-5-iodopyridine-3-carboxylate, 2-Hydroxy-3-iodo-5-(methoxycarbonyl)pyridine
OR110402-1G
OR110402-250MG
OR110402-5G
APOSOR110402-1GEA
870
CZK
APOSOR110402-1G
APOSOR110402-250MG
APOSOR110402-5G
Methyl 6-hydroxy-5-iodonicotinate
Methyl 6-hydroxy-5-iodonicotinate
Vzorec:
C₇H₆INO₃ MW: 279,04 g/mol Bod tání: 203…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD17168292 CAS číslo: 885950-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...