Ethyl-4-hydroxy-3-nitrobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
OR110385-10G
OR110385-5G
APOSOR110385-10GEA
2280
CZK
APOSOR110385-10G
APOSOR110385-5G
Ethyl-4-hydroxy-3-nitrobenzoate
Ethyl-4-hydroxy-3-nitrobenzoate
Vzorec:
C₉H₉NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00016999 CAS číslo: 19013-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...