5-Chloro-6-hydroxynicotinaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR110376-1G
OR110376-100MG
APOSOR110376-1GEA
2400
CZK
APOSOR110376-1G
APOSOR110376-100MG
5-Chloro-6-hydroxynicotinaldehyde
5-Chloro-6-hydroxynicotinaldehyde
Vzorec:
C₆H₄ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04972384 CAS číslo: 627501-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...