3-Nitrobenzhydrazide
Dodavatel: Apollo Scientific
|
Varování
|
OR110308-25G
OR110308-100G
APOSOR110308-25GEA
2440
CZK
APOSOR110308-25G
APOSOR110308-100G
3-Nitrobenzhydrazide
3-Nitrobenzhydrazide
Vzorec:
C₇H₇N₃O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00017059 CAS číslo: 618-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...