3-Pyridineboronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
OR11024-100G
OR11024-25G
OR11024-5G
APOSOR11024-100GEA
4240
CZK
APOSOR11024-100G
APOSOR11024-25G
APOSOR11024-5G
3-Pyridineboronic acid pinacol ester
3-Pyridineboronic acid pinacol ester
Vzorec:
C₁₁H₁₆BNO₂ MW: 205,06 g/mol Bod tání: 102…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD03084758 CAS číslo: 329214-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...