Tisk…

2-Methyl-3-nitrobenzenecarbohydrazide

Dodavatel: Apollo Scientific
Varování

OR110221-10G OR110221-1G OR110221-5G
APOSOR110221-10GEA 12730 CZK
APOSOR110221-10G APOSOR110221-1G APOSOR110221-5G
2-Methyl-3-nitrobenzenecarbohydrazide
2-Methyl-3-nitrobenzenecarbohydrazide
Vzorec: C₈H₉N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD03422927
CAS číslo: 869942-83-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR